accessibility menu, dialog, popup

UserName

Br

CAS RN 14469-83-1

IUPAC Name: (5-bromopentyl)benzene
Synonyms: (5-bromopentyl)benzene
Molecular Weight (g/mol): 227.15
Molecular Formula: C11H15Br
InChi Key: QICUPOFVENZWSC-UHFFFAOYSA-N
SMILES: BrCCCCCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results