missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-5-phenylpentane, 98%

Product Code. 10569223 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10569223 5 g Glass bottle 5g
10564764 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10569223 Supplier Thermo Scientific Acros Supplier No. 384740050
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14469-83-1 | C11H15Br | 227.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14469-83-1
Molecular Formula C11H15Br
Molecular Weight (g/mol) 227.14
MDL Number MFCD01075177
InChI Key QICUPOFVENZWSC-UHFFFAOYSA-N
Synonym 5-bromopentyl benzene, 1-bromo-5-phenylpentane, benzene, 5-bromopentyl, 5-phenylpentyl bromide, 1-bromo-5-phenyl-pentane, 5-bromo-n-pentyl benzene, 5-bromo-pentyl-benzene, 5-brompentyl benzol, phenylpentyl bromide, 5-phenylpentylbromide
PubChem CID 285561
IUPAC Name 5-bromopentylbenzene
SMILES C1=CC=C(C=C1)CCCCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.2300g/mL
Boiling Point 148°C (18.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5(CH2)5Br
Refractive Index 1.5320 to 1.534
Quantity 5 g
Specific Gravity 1.23
Formula Weight 227.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromo-5-phenylpentane
Show More Show Less
compliance-icons
Product Identifier
  • 1-Bromo-5-phenylpentane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.