CAS RN 1449135-41-4
CAS RN 1449135-41-4
IUPAC Name:
tert-butyl N-[4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
Synonyms:
tert-butyl N-[4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
Molecular Weight (g/mol):
337.2
Molecular Formula:
C17H25BFNO4
InChi Key:
JWYMEKZRPHNWGS-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)NC1=CC(B2OC(C)(C)C(C)(C)O2)=C(F)C=C1
1
–
1
of
1
results
1
5-(Boc-amino)-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
CAS: 1449135-41-4 Molecular Formula: C17H25BFNO4 Molecular Weight (g/mol): 337.198 MDL Number: MFCD22419260 InChI Key: JWYMEKZRPHNWGS-UHFFFAOYSA-N Synonym: tert-butyl n-4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate,5-boc-amino-2-fluorobenzeneboronic acid pinacol ester,tert-butyl 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate PubChem CID: 86810446 IUPAC Name: tert-butyl N-[4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate SMILES: B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)NC(=O)OC(C)(C)C)F