accessibility menu, dialog, popup

UserName

CAS RN 1449144-70-0

IUPAC Name: (2-fluoro-5-{[(2-fluorophenyl)methyl]carbamoyl}phenyl)boronic acid
Synonyms: 2-fluoro-5-{[(2-fluorophenyl)methyl]carbamoyl}phenylboronic acid
Molecular Weight (g/mol): 291.06
Molecular Formula: C14H12BF2NO3
InChi Key: KQSVTPWXLNYZIX-UHFFFAOYSA-N
SMILES: OB(O)C1=C(F)C=CC(=C1)C(=O)NCC1=CC=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results