accessibility menu, dialog, popup

UserName

CAS RN 14506-33-3

H2CClOH

CAS RN 14506-33-3

IUPAC Name: 1-(4-chlorophenyl)but-3-en-1-ol
Synonyms: 1-(4-chlorophenyl)but-3-en-1-ol
Molecular Weight (g/mol): 182.65
Molecular Formula: C10H11ClO
InChi Key: GJODRJDLQVBTMF-UHFFFAOYNA-N
SMILES: OC(CC=C)C1=CC=C(Cl)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)