missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-3-buten-1-ol, 97%

Product Code. 15490138
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490138 1 g 1g
15400148 5 g 5g
15410148 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490138 Supplier Thermo Scientific Alfa Aesar Supplier No. H53381.03
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 14506-33-3 | C10H11ClO | 182.647 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14506-33-3
Molecular Formula C10H11ClO
Molecular Weight (g/mol) 182.647
MDL Number MFCD03427246
InChI Key GJODRJDLQVBTMF-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl-3-buten-1-ol, 1-4-chlorophenyl but-3-en-1-ol, benzenemethanol,4-chloro-a-2-propen-1-yl, 4-4-chlorophenyl-1-buten-4-ol, 1-4-chloro-phenyl-but-3-en-1-ol, acmc-20apb1, 4-chloro-alpha-allylbenzyl alcohol, 4-4-chlorophenyl but-1-en-4-ol, 1-4-chlorophenyl-but-3-en-1-ol
PubChem CID 2757771
IUPAC Name 1-(4-chlorophenyl)but-3-en-1-ol
SMILES C=CCC(C1=CC=C(C=C1)Cl)O

Specifications

Density 1.137
Quantity 1 g
Refractive Index 1.544
Boiling Point 140°C (2 mmHg)
Formula Weight 182.65
Percent Purity 97%
Chemical Name or Material 1-(4-Chlorophenyl)-3-buten-1-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.