accessibility menu, dialog, popup

UserName

ONOH

CAS RN 145127-38-4

IUPAC Name: [3-(morpholin-4-yl)phenyl]methanol
Synonyms: [3-(morpholin-4-yl)phenyl]methanol
Molecular Weight (g/mol): 193.25
Molecular Formula: C11H15NO2
InChi Key: MXBVALXTJZMIJB-UHFFFAOYSA-N
SMILES: OCC1=CC(=CC=C1)N1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results