missing translation for 'onlineSavingsMsg'
Learn More

(3-Morpholinophenyl)methanol, 97%, Thermo Scientific™

Product Code. 15249806 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15249806 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15249806 Supplier Thermo Scientific Acros Supplier No. 452590050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 145127-38-4
Melting Point 51.0°C to 56.0°C
Assay Percent Range 96% min. (HPLC)
Molecular Formula C11H15NO2
MDL Number MFCD07772812
Quantity 5 g
Synonym 3-morpholinophenyl methanol, 3-morpholinobenzyl alcohol, 3-morpholin-4-yl phenyl methanol, 3-morpholin-4-ylphenyl methanol, benzenemethanol,3-4-morpholinyl, benzenemethanol, 3-4-morpholinyl, 3-morpholinobenzylalcohol, acmc-20n4f1, 3-morpholin-4-ylphenyl methan-1-ol
InChI Key MXBVALXTJZMIJB-UHFFFAOYSA-N
SMILES OCC1=CC(=CC=C1)N1CCOCC1
IUPAC Name (3-morpholin-4-ylphenyl)methanol
Molecular Weight (g/mol) 193.25
PubChem CID 7162074
Formula Weight 193.24
Percent Purity 97%
Chemical Name or Material (3-Morpholinophenyl)methanol
Show More Show Less
compliance-icons
Product Identifier
  • (3-Morpholinophenyl)methanol, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.