accessibility menu, dialog, popup

UserName

CAS RN 14528-64-4

IUPAC Name: dipotassium 5-nitro-2-(sulfonatooxy)benzen-1-olate dihydrate
Synonyms: dipotassium 5-nitro-2-(sulfonatooxy)benzenolate dihydrate
Molecular Weight (g/mol): 347.38
Molecular Formula: C6H7K2NO9S
InChi Key: ZIAQNWGTQNXFHV-UHFFFAOYSA-L
SMILES: O.O.[K+].[K+].[O-]C1=CC(=CC=C1OS([O-])(=O)=O)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results