accessibility menu, dialog, popup

UserName

CH3SNOHO

CAS RN 14542-16-6

IUPAC Name: 4-methyl-1,3-thiazole-2-carboxylic acid
Synonyms: 4-methyl-1,3-thiazole-2-carboxylic acid
Molecular Weight (g/mol): 143.16
Molecular Formula: C5H5NO2S
InChi Key: GNGDWDFLILPTKL-UHFFFAOYSA-N
SMILES: CC1=CSC(=N1)C(O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results