missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,3-thiazole-2-carboxylic acid, 97%

Product Code. 10738668 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10738668 5 g Glass bottle 5g
10053967 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10738668 Supplier Thermo Scientific Acros Supplier No. 429360050
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 14542-16-6 | C5H5NO2S | 143.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14542-16-6
Molecular Formula C5H5NO2S
Molecular Weight (g/mol) 143.17
MDL Number MFCD07377486
InChI Key GNGDWDFLILPTKL-UHFFFAOYSA-N
Synonym 4-methylthiazole-2-carboxylic acid, 4-methyl-thiazole-2-carboxylic acid, 4-methyl-2-thiazolecarboxylic acid, 2-thiazolecarboxylicacid, 4-methyl, 2-thiazolecarboxylic acid, 4-methyl, acmc-209cuz, 4-methyl-2-carboxythiazole, ksc173q1t, 4-methylthiazole-2-carboxylicacid
PubChem CID 22499336
IUPAC Name 4-methyl-1,3-thiazole-2-carboxylic acid
SMILES CC1=CSC(=N1)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 90.0°C to 92.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 143.17
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-1, 3-thiazole-2-carboxylic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.