accessibility menu, dialog, popup

UserName

NH2HON

CAS RN 14543-43-2

IUPAC Name: 3-amino-4-hydroxybenzonitrile
Synonyms: 3-amino-4-hydroxybenzonitrile
Molecular Weight (g/mol): 134.14
Molecular Formula: C7H6N2O
InChi Key: ZEWCASRNRWXXSO-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1O)C#N
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)