accessibility menu, dialog, popup

UserName

CAS RN 14618-65-6

H2NOSNH2O

CAS RN 14618-65-6

IUPAC Name: 2-[(carbamoylmethyl)sulfanyl]acetamide
Synonyms: thiodiglycolamide
Molecular Weight (g/mol): 148.18
Molecular Formula: C4H8N2O2S
InChi Key: KQNFZEVUCSXNTH-UHFFFAOYSA-N
SMILES: NC(=O)CSCC(N)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results