accessibility menu, dialog, popup

UserName

CAS RN 14667-47-1

CH3OONH2N

CAS RN 14667-47-1

IUPAC Name: methyl 2-aminopyridine-3-carboxylate
Synonyms: methyl 2-aminopyridine-3-carboxylate
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: NZZDEODTCXHCRS-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CN=C1N
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (2)