missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-aminonicotinate, 97%, Thermo Scientific™

Product Code. 10544885
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10544885 25 g Amber Glass Bottle 25g
10337594 1 g Amber Glass Bottle 1g
10525574 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10544885 Supplier Thermo Scientific Maybridge Supplier No. KM07908EB
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 14667-47-1
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00128872
InChI Key NZZDEODTCXHCRS-UHFFFAOYSA-N
PubChem CID 351633
IUPAC Name methyl 2-aminopyridine-3-carboxylate
SMILES COC(=O)C1=CC=CN=C1N

Specifications

Packaging Amber Glass Bottle
Quantity 25 g
Synonym methyl 2-aminonicotinate, 2-amino-nicotinic acid methyl ester, 2-aminonicotinic acid methyl ester, 3-pyridinecarboxylic acid, 2-amino-, methyl ester, methyl2-aminonicotinate, pubchem16778, methyl?2-aminonicotinate, methyl 2-amino-nicotinate, acmc-1c0pk, aminonicotinsauremethylester
Formula Weight 152.15
Percent Purity 97%
Chemical Name or Material methyl 2-aminonicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.