accessibility menu, dialog, popup

UserName

H3CCl

CAS RN 1467-05-6

IUPAC Name: 1-(chloromethyl)-4-ethylbenzene
Synonyms: 1-(chloromethyl)-4-ethylbenzene
Molecular Weight (g/mol): 154.64
Molecular Formula: C9H11Cl
InChi Key: DUBCVXSYZVTCOC-UHFFFAOYSA-N
SMILES: CCC1=CC=C(CCl)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results