accessibility menu, dialog, popup

UserName

H3CCH3OOOCH3CH3

CAS RN 1468-39-9

IUPAC Name: 3-methylbutanoyl 3-methylbutanoate
Synonyms: 3-methylbutanoyl 3-methylbutanoate
Molecular Weight (g/mol): 186.25
Molecular Formula: C10H18O3
InChi Key: FREZLSIGWNCSOQ-UHFFFAOYSA-N
SMILES: CC(C)CC(=O)OC(=O)CC(C)C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)