missing translation for 'onlineSavingsMsg'
Learn More

Isovaleric anhydride, 95%, Thermo Scientific Chemicals

Product Code. 15254864 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15254864 100 mL Glass bottle 100mL
15234864 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15254864 Supplier Thermo Scientific Acros Supplier No. 451091000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1468-39-9
Molecular Formula C10H18O3
Molecular Weight (g/mol) 186.25
MDL Number MFCD00015051
InChI Key FREZLSIGWNCSOQ-UHFFFAOYSA-N
Synonym isovaleric anhydride, 3-methylbutanoic anhydride, butanoic acid, 3-methyl-, anhydride, butanoic acid, 3-methyl-, 1,1'-anhydride, isovalericanhydride, isovaleriansaureanhydrid, iso-pentanoic anhydride, acmc-209cxm, isovaleric anhydride gc, butanoic acid,3-methyl-,1,1'-anhydride
PubChem CID 73847
IUPAC Name 3-methylbutanoyl 3-methylbutanoate
SMILES CC(C)CC(=O)OC(=O)CC(C)C

Specifications

Density 0.9400g/mL
Boiling Point 215°C
Flash Point 91°C
Assay Percent Range 94% min
Packaging Glass bottle
Quantity 100 mL
Specific Gravity 0.94
Solubility Information Solubility in water: hydrolyses
Formula Weight 186.25
Percent Purity 95%
Chemical Name or Material Isovaleric anhydride
Show More Show Less
compliance-icons
Product Identifier
  • Isovaleric anhydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P390-Absorb spillage to prevent material damage.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.