accessibility menu, dialog, popup

UserName

OHFBr

CAS RN 147460-41-1

IUPAC Name: 2-bromo-5-fluorophenol
Synonyms: 2-bromo-5-fluorophenol
Molecular Weight (g/mol): 191
Molecular Formula: C6H4BrFO
InChi Key: HUVAOAVBKOVPBZ-UHFFFAOYSA-N
SMILES: OC1=CC(F)=CC=C1Br
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)