accessibility menu, dialog, popup

UserName

OHBrOHBr

CAS RN 14753-51-6

IUPAC Name: 2,5-dibromobenzene-1,4-diol
Synonyms: 2,5-dibromobenzene-1,4-diol
Molecular Weight (g/mol): 267.9
Molecular Formula: C6H4Br2O2
InChi Key: VALXCIRMSIFPFN-UHFFFAOYSA-N
SMILES: OC1=CC(Br)=C(O)C=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results