CAS RN 148-75-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Hydroxy-2,7-naphthalenedisulfonic Acid Sodium Salt >90%, TRC
CAS: 148-75-4 Molecular Formula: C10 H8 O7 S2 Molecular Weight (g/mol): 304.3 Synonym: 2-Hydroxy-3,6-naphthalenedisulfonic Acid Sodium Salt,2-Naphthol-3,6-disulfonic Acid Sodium Salt,Acid Chrome Black V Sodium Salt,Magracrom Black V Sodium Salt,R Acid Sodium Salt,β-Naphthol-3,6-disulfonic Acid Sodium Salt,β-Naphtholdisulfonic Acid R Sodium Salt IUPAC Name: 3-hydroxynaphthalene-2,7-disulfonic acid SMILES: Oc1cc2ccc(cc2cc1S(=O)(=O)O)S(=O)(=O)O