accessibility menu, dialog, popup

UserName

CH3BrOCH3

CAS RN 14804-31-0

IUPAC Name: 4-bromo-1-methoxy-2-methylbenzene
Synonyms: 4-bromo-1-methoxy-2-methylbenzene
Molecular Weight (g/mol): 201.06
Molecular Formula: C8H9BrO
InChi Key: UDLRGQOHGYWLCS-UHFFFAOYSA-N
SMILES: COC1=C(C)C=C(Br)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results