CAS RN 1482-50-4
CAS RN 1482-50-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5beta-Dihydrocortisol, TRC
CAS: 1482-50-4 Molecular Formula: C21 H32 O5 Molecular Weight (g/mol): 364.48 Synonym: Pregnane-3,20-dione, 11,17,21-trihydroxy-, (5β,11β)-,5β-Pregnane-3,20-dione, 11β,17,21-trihydroxy- (7CI,8CI),(5β,11β)-11,17,21-Trihydroxypregnane-3,20-dione,11β,17,21-Trihydroxy-5β-pregnane-3,20-dione,11β,17α,21-Trihydroxy-5β-pregnane-3,20-dione,5β-Dihydrocortisol,Cortisol, dihydro-,Dihydrocortisol,NSC 15473 IUPAC Name: (5R,8S,9S,10S,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,4,5,6,7,8,9,11,12,14,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-3-one SMILES: C[C@]12CCC(=O)C[C@H]1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)CO)[C@@]4(C)C[C@H](O)[C@H]23