accessibility menu, dialog, popup

UserName

CH3SOOHNBOHHO

CAS RN 148355-75-3

IUPAC Name: (3-methanesulfonamidophenyl)boronic acid
Synonyms: 3-methanesulfonamidophenylboronic acid
Molecular Weight (g/mol): 215.03
Molecular Formula: C7H10BNO4S
InChi Key: XUIQQIRLFMCWLN-UHFFFAOYSA-N
SMILES: CS(=O)(=O)NC1=CC=CC(=C1)B(O)O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)