accessibility menu, dialog, popup

UserName

CH3OONNCl

CAS RN 149849-94-5

IUPAC Name: methyl 2-chloropyrimidine-4-carboxylate
Synonyms: methyl 2-chloropyrimidine-4-carboxylate
Molecular Weight (g/mol): 172.57
Molecular Formula: C6H5ClN2O2
InChi Key: GGTNGWOGJHJQCL-UHFFFAOYSA-N
SMILES: COC(=O)C1=NC(Cl)=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results