missing translation for 'onlineSavingsMsg'
Learn More

methyle2-chloropyrimidine-4-carboxylate, 97%, Thermo Scientific™

Product Code. 10662305
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10662305 250 mg Amber Glass Bottle 250mg
10442464 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10662305 Supplier Thermo Scientific Maybridge Supplier No. MO08502CB
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 149849-94-5
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.57
MDL Number MFCD11841080
InChI Key GGTNGWOGJHJQCL-UHFFFAOYSA-N
Synonym 2-chloro-pyrimidine-4-carboxylic acid methyl ester, 2-chloropyrimidine-4-carboxylic acid methyl ester, 4-pyrimidinecarboxylic acid, 2-chloro-, methyl ester, 2-chloro-4-methoxycarbonyl pyrimidine, 2-chloro-4-pyrimidinecarboxylic acid methyl ester, 4-pyrimidinecarboxylicacid, 2-chloro-, methyl ester, methyl2-chloropyrimidine-4-carboxylate
PubChem CID 43811076
IUPAC Name methyl 2-chloropyrimidine-4-carboxylate
SMILES COC(=O)C1=NC(Cl)=NC=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 172.57
Percent Purity 97%
Chemical Name or Material Methyl 2-chloropyrimidine-4-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.