accessibility menu, dialog, popup

UserName

H3CCH3(R)CH3(R)CH3CH3OH(E)

CAS RN 150-86-7

IUPAC Name: (2E,7R,11R)-3,7,11,15-tetramethylhexadec-2-en-1-ol
Synonyms: phytol
Molecular Weight (g/mol): 296.54
Molecular Formula: C20H40O
InChi Key: BOTWFXYSPFMFNR-PYDDKJGSSA-N
SMILES: CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC\C(C)=C\CO
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results