missing translation for 'onlineSavingsMsg'
Learn More

trans-Phytol, TRC

Product Code. 30136141
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136141 5 g 5g
30134071 2.5 g 2.5g
30010629 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136141 Supplier Toronto Research Chemicals Supplier No. TRCP3989105G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-86-7
Molecular Formula C20H40O
Molecular Weight (g/mol) 296.53
Synonym [R-[R*,R*-(E)]]- 3,7,11,15-tetramethyl-2-hexadecen-1-ol, Phytol (6CI,8CI), (2E,7R,11R)-3,7,11,15-Tetramethyl-2-hexadecen-1-ol, (7R,11R,2E)-Phytol;(E)-Phytol, (E,R,R)-Phytol, 3,7,11,15-Tetramethylhexadec-2-en-1-ol
IUPAC Name (E,7R,11R)-3,7,11,15-tetramethylhexadec-2-en-1-ol
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC\C(=C\CO)\C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C20H40O/c1-17(2)9-6-10-18(3)11-7-12-19(4)13-8-14-20(5)15-16-21/h15,17-19,21H,6-14,16H2,1-5H3/b20-15+/t18-,19-/m1/s1
Formula Weight 296.3079
Chemical Name or Material trans-Phytol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.