accessibility menu, dialog, popup

UserName

NH4NH4SHSHSMoS

CAS RN 15060-55-6

IUPAC Name: diammonium disulfanylidenemolybdenum disulfanide
Synonyms: ammonium tetrathiomolybdate
Molecular Weight (g/mol): 262.28
Molecular Formula: H10MoN2S4
InChi Key: ZKKLPDLKUGTPME-UHFFFAOYSA-N
SMILES: [NH4+].[NH4+].[SH-].[SH-].S=[Mo]=S
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (1)
  • (6)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
  • (4)