missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Tetrathiomolybdate, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | Ammonium Tetrathiomolybdate |
| CAS | 15060-55-6 |
| Molecular Formula | MoS4 . 2(NH4) |
| Quantity | 2.5 g |
| Synonym | ATTM, Ammonium Molybdenum Sulfide, Ammonium Tetrasulfidomolybdate(2-), Ammonium Thiomolybdate, Bis(ammonium)tetrathiomolybdate(2-), Coprexa, Thiomolybdic Acid Diammonium Salt, Diammonium Tetrakis(sulfido)molybdate(2-), Diammonium Tetrakis(thioxo)molybdate, Diammonium Tetrasulfidomolybdate, Diammonium Tetrathiomolybdate, Diammonium Tetrathiomolybdate(2-), Diammonium Tetrathiooxomolybdate(2-), Diammonium Tetrathioxomolybdate(2-), Diammonium Thiomolybdate, Diammonium Thiomolybdate, NSC 286644, NSC 714598, TM, Tiomolibdate Diammonium |
| InChI Formula | InChI=1S/Mo.2H3N.4S/h;2*1H3;;;;/q;;;;;2*-1/p+2 |
| SMILES | [NH4+].[NH4+].[S-][Mo](=S)(=S)[S-] |
| IUPAC Name | diazanium;bis(sulfanylidene)-disulfidomolybdenum |
| Molecular Weight (g/mol) | 262.3 |
| Formula Weight | 261.8624 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto