accessibility menu, dialog, popup

UserName

CAS RN 151504-80-2

NOOClFFF

CAS RN 151504-80-2

IUPAC Name: 2-chloro-4-nitro-1-(trifluoromethyl)benzene
Synonyms: 2-chloro-4-nitro-1-(trifluoromethyl)benzene
Molecular Weight (g/mol): 225.55
Molecular Formula: C7H3ClF3NO2
InChi Key: MOTWHSMEZCFDOD-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results