CAS RN 151882-81-4
CAS RN 151882-81-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
BTUM, TRC
CAS: 151882-81-4 Molecular Formula: C29 H28 N4 O6 S2 Molecular Weight (g/mol): 592.69 Synonym: N,N'-[Methylenebis(4,1-phenyleneiminocarbonyl)]bis[4-methylbenzenesulfonamide],4,4'-Bis(p-toluenesulfonylaminocarbonylamino)diphenylmethane,4,4'-Bis(p-toluenesulfonylaminocarboxylamino)diphenylmethane,4,4'-Bis(p-tolylsulfonylureido)diphenylmethane,BTUM IUPAC Name: 1-(4-methylphenyl)sulfonyl-3-[4-[[4-[(4-methylphenyl)sulfonylcarbamoylamino]phenyl]methyl]phenyl]urea SMILES: Cc1ccc(cc1)S(=O)(=O)NC(=O)Nc2ccc(Cc3ccc(NC(=O)NS(=O)(=O)c4ccc(C)cc4)cc3)cc2