CAS RN 152503-50-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Diosmetin 3-O-Beta-D-Glucuronide, TRC
CAS: 152503-50-9 Molecular Formula: C22 H20 O12 Molecular Weight (g/mol): 476.39 Synonym: 5-(5,7-Dihydroxy-4-oxo-4H-1-benzopyran-2-yl)-2-methoxyphenyl-β-D-Glucopyranosiduronic Acid IUPAC Name: (2S,3S,4S,5R,6S)-6-[5-(5,7-dihydroxy-4-oxochromen-2-yl)-2-methoxyphenoxy]-3,4,5-trihydroxyoxane-2-carboxylic acid SMILES: COc1ccc(cc1O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O)C3=CC(=O)c4c(O)cc(O)cc4O3