CAS RN 15299-99-7
CAS RN 15299-99-7
IUPAC Name:
N,N-diethyl-2-(naphthalen-1-yloxy)propanamide
Synonyms:
N,N-diethyl-2-(naphthalen-1-yloxy)propanamide
Molecular Weight (g/mol):
271.36
Molecular Formula:
C17H21NO2
InChi Key:
WXZVAROIGSFCFJ-UHFFFAOYNA-N
SMILES:
CCN(CC)C(=O)C(C)OC1=C2C=CC=CC2=CC=C1
1
–
2
of
2
results
1
Napropamid, TRC
CAS: 15299-99-7 Molecular Formula: C17 H21 N O2 Molecular Weight (g/mol): 271.35 Synonym: Propionamide, N,N-diethyl-2-(1-naphthyloxy)- (8CI),N,N-Diethyl-2-(1-naphthalenyloxy)propanamide,(±)-Devrinol,2-(α-Naphthoxy)-N,N-diethylpropionamide,Devrinol,N,N-Diethyl-2-(1-naphthalenyloxy)propionamide,N,N-Diethyl-2-(1-naphthyloxy)propanamide,Napropamid,Napropamide,R 7465,Racemic devrinol IUPAC Name: N,N-diethyl-2-naphthalen-1-yloxypropanamide SMILES: CCN(CC)C(=O)C(C)Oc1cccc2ccccc12
2
Napropamide, MedChemExpress
MedChemExpress Napropamide is a selective systemic amide herbicide used to control a number of annual grasses and broad-leaved weeds.