accessibility menu, dialog, popup

UserName

H3COHNNHOCH3

CAS RN 1530-73-0

IUPAC Name: 4-methyl-N'-(4-methylbenzoyl)benzohydrazide
Synonyms: 4-methyl-N'-(4-methylbenzoyl)benzohydrazide
Molecular Weight (g/mol): 268.32
Molecular Formula: C16H16N2O2
InChi Key: SWVLEZBOJRWRJB-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(=O)NNC(=O)C1=CC=C(C)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results