accessibility menu, dialog, popup

UserName

BrN

CAS RN 1532-97-4

IUPAC Name: 4-bromoisoquinoline
Synonyms: 4-bromoisoquinoline
Molecular Weight (g/mol): 208.06
Molecular Formula: C9H6BrN
InChi Key: SCRBSGZBTHKAHU-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC=CC2=CN=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (3)
  • (5)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)