missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoisoquinoline, 98%

Product Code. 10559810 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10559810 1 g Glass bottle 1g
10487090 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10559810 Supplier Thermo Scientific Acros Supplier No. 107090010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1532-97-4 | C9H6BrN | 208.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1532-97-4
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.06
MDL Number MFCD00006904
InChI Key SCRBSGZBTHKAHU-UHFFFAOYSA-N
Synonym isoquinoline, 4-bromo, 4-bromo-isoquinoline, 4-isoquinolinyl bromide, 4-bromisochinolin, 4bromoisoquinoline, 4-bromoisoquinoline, 4-bromo isoquinoline, 4-bromoisoquinoline?, pubchem6212, acmc-1bu1h
PubChem CID 73743
IUPAC Name 4-bromoisoquinoline
SMILES BrC1=C2C=CC=CC2=CN=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 39.0°C to 43.0°C
Color White to Beige
Boiling Point 280.0°C to 285.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Beilstein 20, 384
Formula Weight 208.06
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 4-Bromoisoquinoline, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromoisoquinoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.