accessibility menu, dialog, popup

UserName

HOBOHFFFFFF

CAS RN 153254-09-2

IUPAC Name: [2,4-bis(trifluoromethyl)phenyl]boronic acid
Synonyms: 2,4-bis(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol): 257.93
Molecular Formula: C8H5BF6O2
InChi Key: WLYPBMBWKYALCG-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results