accessibility menu, dialog, popup

UserName

H3CO

CAS RN 1533-20-6

IUPAC Name: 1,3-diphenylbutan-1-one
Synonyms: 1,3-diphenylbutan-1-one
Molecular Weight (g/mol): 224.3
Molecular Formula: C16H16O
InChi Key: GIVFXLVPKFXTCU-UHFFFAOYNA-N
SMILES: CC(CC(=O)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results