CAS RN 154492-74-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Phenoxy-d5-acetic Acid, TRC
CAS: 154492-74-7 Molecular Formula: C8 2H5 H3 O3 Molecular Weight (g/mol): 157.18 Synonym: Acetic acid, phenoxy-d5- (9CI),Phenoxy-d5-acetic acid,Phenoxyacetic acid D5 (phenoxy-D5),2-Phenoxy-d5-acetic acid,2-(Phenyl-d5-oxy)acetic acid,Glycolic acid phenyl-d5 ether,Phenoxyacetic acid-d5,Pentadeuteriophenoxyethanoic acid IUPAC Name: 2-(2,3,4,5,6-pentadeuteriophenoxy)acetic acid SMILES: [2H]c1c([2H])c([2H])c(OCC(=O)O)c([2H])c1[2H]
2
Phenoxy-d5-acetic Acid, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.