CAS RN 15469-38-2
CAS RN 15469-38-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Iron(III) fluoride trihydrate
CAS: 15469-38-2 MDL Number: MFCD00149718 Synonym: iron 3+ trifluoride trihydrate,acmc-20akpl,ferric iron trihydrate trifluoride,iron 3+ ion trihydrate trifluoride,iron 3+ fluoride-water 1/3/3,fluoride, fluoride, iron salt, hydrate, hydrate, hydrate, fluoride