accessibility menu, dialog, popup

UserName

CAS RN 156022-15-0

CH3FOH

CAS RN 156022-15-0

IUPAC Name: 1-(2-fluorophenyl)propan-1-ol
Synonyms: 1-(2-fluorophenyl)propan-1-ol
Molecular Weight (g/mol): 154.18
Molecular Formula: C9H11FO
InChi Key: MCENMZBHIHBHPB-UHFFFAOYNA-N
SMILES: CCC(O)C1=CC=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results