CAS RN 1561-11-1
CAS RN 1561-11-1
IUPAC Name:
4-methylhexanoic acid
Synonyms:
4-methylhexanoic acid
Molecular Weight (g/mol):
130.19
Molecular Formula:
C7H14O2
InChi Key:
DIVCBWJKVSFZKJ-UHFFFAOYNA-N
SMILES:
CCC(C)CCC(O)=O
1
–
1
of
1
results
1
4-Methylhexanoic Acid, TRC
CAS: 1561-11-1 Molecular Formula: C7H14O2 Molecular Weight (g/mol): 130.18 Synonym: (±)-4-Methylhexanoic Acid,4-Ethylpentanoic Acid,4-Methylcaproic Acid,NSC 249817 IUPAC Name: 4-methylhexanoic acid SMILES: CCC(C)CCC(=O)O