missing translation for 'onlineSavingsMsg'
Learn More

4-Methylhexanoic Acid, TRC

Product Code. 30478588
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478588 100 mg 100mg
30476415 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30478588 Supplier Toronto Research Chemicals Supplier No. TRCM312403100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1561-11-1
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.18
Synonym (±)-4-Methylhexanoic Acid, 4-Ethylpentanoic Acid, 4-Methylcaproic Acid, NSC 249817
IUPAC Name 4-methylhexanoic acid
SMILES CCC(C)CCC(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C7H14O2/c1-3-6(2)4-5-7(8)9/h6H,3-5H2,1-2H3,(H,8,9)
Formula Weight 130.0994
Chemical Name or Material 4-Methylhexanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.