accessibility menu, dialog, popup

UserName

HOBOHFF

CAS RN 156545-07-2

IUPAC Name: (3,5-difluorophenyl)boronic acid
Synonyms: 3,5-difluorophenylboronic acid
Molecular Weight (g/mol): 157.91
Molecular Formula: C6H5BF2O2
InChi Key: QWQBQRYFWNIDOC-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(F)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results