accessibility menu, dialog, popup

UserName

CH3OOOOH

CAS RN 157101-77-4

IUPAC Name: 7-hydroxy-4-(methoxymethyl)-2H-chromen-2-one
Synonyms: 7-hydroxy-4-(methoxymethyl)chromen-2-one
Molecular Weight (g/mol): 206.2
Molecular Formula: C11H10O4
InChi Key: PRVOZGKAZHXKMM-UHFFFAOYSA-N
SMILES: COCC1=CC(=O)OC2=CC(O)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results