CAS RN 1572-98-1
IUPAC Name:
ethyl 2-cyano-2,2-dimethylacetate
Synonyms:
ethyl 2-cyano-2,2-dimethylacetate
Molecular Weight (g/mol):
141.17
Molecular Formula:
C7H11NO2
InChi Key:
FYGRPGOHQCPZCV-UHFFFAOYSA-N
SMILES:
CCOC(=O)C(C)(C)C#N
1
–
1
of
1
results
1
Ethyl 2-cyano-2-methylpropionate, 97%
CAS: 1572-98-1 Molecular Formula: C7H11NO2 Molecular Weight (g/mol): 141.17 MDL Number: MFCD00034667 InChI Key: FYGRPGOHQCPZCV-UHFFFAOYSA-N PubChem CID: 344887 IUPAC Name: ethyl 2-cyano-2-methylpropanoate SMILES: CCOC(=O)C(C)(C)C#N