missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-cyano-2-methylpropionate, 97%

Product Code. 15444087
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15444087 50 g 50g
15434087 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15444087 Supplier Thermo Scientific Alfa Aesar Supplier No. H31432.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1572-98-1 | C7H11NO2 | 141.17 g/mol

It is employed as a intermediates for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediates for pharmaceutical and organic synthesis.

Solubility
Soluble in ether. Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1572-98-1
Molecular Formula C7H11NO2
Molecular Weight (g/mol) 141.17
MDL Number MFCD00034667
InChI Key FYGRPGOHQCPZCV-UHFFFAOYSA-N
PubChem CID 344887
IUPAC Name ethyl 2-cyano-2-methylpropanoate
SMILES CCOC(=O)C(C)(C)C#N

Specifications

Density 0.97
Boiling Point 185°C
Quantity 50 g
Synonym 2-cyano-2-methylpropionic acid ethyl ester, ethyl 2-cyano-2-methylpropionate, ethyl 2-cyano-2,2-dimethylacetate, 2-cyano-2-methylpropanoic acid ethyl ester, ethyl 2-cyano-2-methyl-propanoate, propanoic acid, 2-cyano-2-methyl-, ethyl ester, pubchem23352, acmc-209dfv, ksc529g3h, ethyl2-cyano-2-methylpropionate
Solubility Information Soluble in ether. Not miscible or difficult to mix with water.
Formula Weight 141.17
Percent Purity 97%
Chemical Name or Material Ethyl 2-cyano-2-methylpropionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.