CAS RN 1583289-85-3
CAS RN 1583289-85-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
p-SCN-Bn-NOTA Trihydrochloride, Technical Grade, TRC
CAS: 1583289-85-3 Molecular Formula: C20H26N4O6S . 3HCl Molecular Weight (g/mol): 559.9 Synonym: 2-S-(4-Isothiocyanatobenzyl)-1,4,7-triazacyclononane-1,4,7-triacetic Acid Trihydrochloride,2-(4-Isothiocyanatobenzyl)-1,4,7-triazacyclonane-1,4,7-triacetic Acid Trihydrochloride IUPAC Name: 2-[4,7-bis(carboxymethyl)-5-[(4-isothiocyanatophenyl)methyl]-1,4,7-triazonan-1-yl]acetic acid;trihydrochloride SMILES: Cl.Cl.Cl.OC(=O)CN1CCN(CC(=O)O)CC(Cc2ccc(cc2)N=C=S)N(CC(=O)O)CC1