missing translation for 'onlineSavingsMsg'
Learn More

p-SCN-Bn-NOTA Trihydrochloride, Technical Grade, TRC

Product Code. 30422388
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30422388 1 mg
30418315 10 mg
30421147 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30422388 Supplier Toronto Research Chemicals Supplier No. TRCS2001201MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1583289-85-3
Molecular Formula C20H26N4O6S . 3HCl
Molecular Weight (g/mol) 559.9
Synonym 2-S-(4-Isothiocyanatobenzyl)-1,4,7-triazacyclononane-1,4,7-triacetic Acid Trihydrochloride, 2-(4-Isothiocyanatobenzyl)-1,4,7-triazacyclonane-1,4,7-triacetic Acid Trihydrochloride
IUPAC Name 2-[4,7-bis(carboxymethyl)-5-[(4-isothiocyanatophenyl)methyl]-1,4,7-triazonan-1-yl]acetic acid;trihydrochloride
SMILES Cl.Cl.Cl.OC(=O)CN1CCN(CC(=O)O)CC(Cc2ccc(cc2)N=C=S)N(CC(=O)O)CC1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C20H26N4O6S.3ClH/c25-18(26)11-22-5-6-23(12-19(27)28)10-17(24(8-7-22)13-20(29)30)9-15-1-3-16(4-2-15)21-14-31;;;/h1-4,17H,5-13H2,(H,25,26)(H,27,28)(H,29,30);3*1H
Formula Weight 558.0873
Chemical Name or Material p-SCN-Bn-NOTA Trihydrochloride, Technical Grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.